ContactUsLinkComponent
Media bottle closures, Top Works™ system, Nalgene®
Media bottle closures, Top Works™ system, Nalgene®
Catalog # 700-0001
   Low Manufacturing Impact
undefined
Media bottle closures, Top Works™ system, Nalgene®
Catalog # 700-0001
   Low Manufacturing Impact
Specifications
  • Connection:
    2x6,35 mm/1x3,2 mm
  • Thread:
    GL 45
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Pk:
    1
  • Environmental certification:
    NAR Certificate of Retirement
Specifications
About this item
PSF, closures with platinum cured silicone inserts.
  • They fit the corresponding threads on bottles by Schott, Corning and Wheaton
  • For thread size GL 45
  • Autoclavable, leakproof
These closures make it easier to transfer liquids aseptically from most conventional glass media bottles.