Orders LinkContactUsLinkComponent
Hook connectors
Hook connectors
Supplier:   USBECK Laborgeräte
Hook connectors
Certificates
Hook connectors
3 Options Available Below
Product Details & Documents
Nickel plated brass screw with plastic handle.
  • For rods up to 13 mm
  • Angle: 90°
Product Family Options
Product Information
  • Material
    For rod Ø
    Availability
    Price
  • USBE2430NEU
    Hook connector
    Hook connector
    Stainless steel
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,80 €
    19,80 €
    1 * 1 ST
       19,80 €
  • 241-7242
    Hook connector
    Hook connector
    Nickel coated steel
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,20 €
    12,20 €
    1 * 1 ST
       12,20 €
  • 241-0382
    Hook connector
    Hook connector
    Steel, powder coated
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,20 €
    12,20 €
    1 * 1 ST
       12,20 €