Measuring cylinders, conical, Nalgene®
Supplier: Thermo Fisher Scientific
Measuring cylinders, conical, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PMP, transparent.
- Excellent chemical resistance
- Unbreakable, autoclavable
- The liquid volume can be read easily because no meniscus is formed
Ideal for pharmaceutical applications. Graduations in ml and oz. With octagonal foot for improved stability, and spout.
Certifications: Conforms to the volume and accuracy requirements of ASTM E1094.
4 Options Available Below
Product Details & Documents
PMP, transparent.
- Excellent chemical resistance
- Unbreakable, autoclavable
- The liquid volume can be read easily because no meniscus is formed
Ideal for pharmaceutical applications. Graduations in ml and oz. With octagonal foot for improved stability, and spout.
Certifications: Conforms to the volume and accuracy requirements of ASTM E1094.
Specifications for Product Family
Specifications
- Catalog No:
- NALG3673-0004
- 213-0259
- 213-0260
- 213-0261
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Family Options
Product Information
- CapacityToleranceAvailabilityPrice
- Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.NALG3673-0004Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.213-0259Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.213-0260Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.213-0261Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact