ContactUsLinkComponent
VWR®, Measuring Beakers with Handle
VWR®, Measuring Beakers with Handle
Catalog # 213-3403
Supplier:  VWR Collection
Supplier #:
undefined
VWR®, Measuring Beakers with Handle
Catalog # 213-3403
Supplier:  VWR Collection
Supplier #:
Log in for availability
Select an Option
116 mm
Select an Option
20 ml
Select an Option
1000 ml
Select an Option
182 mm
Select an Option
03
Select an Option
1
About this item
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning
  • With embossed scale, blue graduations and spout
Ideal for general laboratory use and a wide variety of other applications.
Certifications: Food-safe products acc. to EC Directive No. 10/2011.
Frequently Bought Together