ContactUsLinkComponent
Beakers, large volume, PP, SP Bel-Art
Beakers, large volume, PP, SP Bel-Art
Catalog # 213-0007
Supplier #: F262190010
undefined
Beakers, large volume, PP, SP Bel-Art
Catalog # 213-0007
Supplier #: F262190010
Log in for availability
Select an Option
269 mm
Select an Option
200 ml
Select an Option
10000 ml
Select an Option
320 mm
Select an Option
02
Select an Option
1
About this item
PP, translucent.
  • Good chemical resistance
  • Steam autoclavable at 121 °C (250 °F)
  • Smooth surfaces for easy, fast cleaning
  • Flared rims and spouts for quick, easy pouring when large volumes are handled
5 liter beakers are graduated every 100 ml, 10 liter beakers are graduated every 200 ml.
Frequently Bought Together