Orders LinkContactUsLinkComponent
Beakers, low form
Beakers, low form
Supplier:   VITLAB
Beakers, low form
Supplier:  VITLAB
Certificates
Beakers, low form
Beakers, low form
6 Options Available Below
Product Details & Documents
PFA, translucent, with spout.
  • Excellent chemical resistance
  • High thermal resistance from –270 to +250 °C
  • Autoclavable
Ideal for demanding applications. Not for use on hotplates. With engraved graduation.
ISO 7056
Certifications: ISO 7056
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Graduation Interval
    Availability
    Price
  • 25 ml
    50 mm
    32 mm
    5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,00 €
    33,00 €
    1 * 1 ST
       33,00 €
  • 50 ml
    59 mm
    39 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,60 €
    41,60 €
    1 * 1 ST
       41,60 €
  • 100 ml
    72 mm
    50 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,00 €
    58,00 €
    1 * 1 ST
       58,00 €
  • 250 ml
    96 mm
    67 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 97,70 €
    97,70 €
    1 * 1 ST
       97,70 €
  • 500 ml
    122 mm
    88 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 152,00 €
    152,00 €
    1 * 1 ST
       152,00 €
  • 1000 ml
    141 mm
    109 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 269,00 €
    269,00 €
    1 * 1 ST
       269,00 €