Orders LinkContactUsLinkComponent
Beakers, low form
Beakers, low form
Supplier:   VITLAB
Beakers, low form
Supplier:  VITLAB
Certificates
Beakers, low form
Beakers, low form
13 Options Available Below
Product Details & Documents
PP, highly transparent.
  • Good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and exceptionally easy-to-read embossed blue graduations.
Food-safe products acc. to EC Directive No. 10/2011.
 : ISO 7056 - 10 ml/150 ml/400 ml/600 ml/3000 ml in addition to ISO 7056)
Product Family Options
Printed graduations, not raised
Product Information
  • Capacity
    Height
    Ext.Ø
    Graduation Interval
    Availability
    Price
  • 10 ml
    36 mm
    30 mm
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,00 €
    2,00 €
    1 * 1 ST
       2,00 €
  • 25 ml
    50 mm
    38 mm
    5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,00 €
    2,00 €
    1 * 1 ST
       2,00 €
  • 50 ml
    60 mm
    47 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,30 €
    2,30 €
    1 * 1 ST
       2,30 €
Embossed graduations
Product Information
  • Capacity
    Height
    Ext.Ø
    Graduation Interval
    Availability
    Price
  • 100 ml
    70 mm
    55 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,40 €
    2,40 €
    1 * 1 ST
       2,40 €
  • 150 ml
    80 mm
    66 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,00 €
    3,00 €
    1 * 1 ST
       3,00 €
  • 250 ml
    95 mm
    77 mm
    25 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,40 €
    3,40 €
    1 * 1 ST
       3,40 €
  • 400 ml
    111,5 mm
    87 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,10 €
    5,10 €
    1 * 1 ST
       5,10 €
  • 500 ml
    117,5 mm
    94 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,90 €
    5,90 €
    1 * 1 ST
       5,90 €
  • 600 ml
    126,5 mm
    100 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,30 €
    6,30 €
    1 * 1 ST
       6,30 €
  • 1000 ml
    147 mm
    120 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,60 €
    7,60 €
    1 * 1 ST
       7,60 €
  • 2000 ml
    187 mm
    149 mm
    200 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,20 €
    13,20 €
    1 * 1 ST
       13,20 €
  • 3000 ml
    212 mm
    170 mm
    250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,60 €
    21,60 €
    1 * 1 ST
       21,60 €
  • 5000 ml
    247 mm
    203 mm
    500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,80 €
    28,80 €
    1 * 1 ST
       28,80 €