Beakers, low form, Nalgene®
Supplier: Thermo Fisher Scientific
Beakers, low form, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PTFE/PFA, transparent, without spout.
- Excellent chemical resistance
- High thermal resistance from –270 to +250 °C
- Autoclavable
Ideal for demanding applications. Not for use on hotplates. With engraved graduation. No drip, single spout design provides precision pouring control.
5 Options Available Below
Product Details & Documents
PTFE/PFA, transparent, without spout.
- Excellent chemical resistance
- High thermal resistance from –270 to +250 °C
- Autoclavable
Ideal for demanding applications. Not for use on hotplates. With engraved graduation. No drip, single spout design provides precision pouring control.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 213-0212
- 213-7212
- 213-7214
- 213-7216
- 213-7217
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
- CapacityAvailabilityPrice
- Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.213-0212Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.213-7212Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.213-7214Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.213-7216Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.213-7217Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact