Orders LinkContactUsLinkComponent
Beakers, low form, Nalgene®
Beakers, low form, Nalgene®
   Sustainable Options Available
Beakers, low form, Nalgene®
   Sustainable Options Available
Certificates
Beakers, low form, Nalgene®
9 Options Available Below
Product Details & Documents
PMP (TPX) crystal clear.
  • Good chemical resistance
  • Heat resistant up to +150 °C, autoclavable at +121 °C (max. 20 minutes)
  • Smooth surfaces for easy, fast cleaning
Designed for general laboratory use. With spout and easy to read, printed graduations with size code, plastic code, maximum temperature for use and 'Not flameproof' symbol. Ribbed for easy stacking. No drip, single spout design provides precision pouring control.
ISO 7056
Certifications: ISO 7056
Specifications for Product Family
Specifications
  • Catalog No:
  • 213-7130
  • 213-7131
  • 213-7132
  • 213-7133
  • 213-7134
  • 213-7135
  • 213-7136
  • 213-7137
  • 213-7138
  • 213-7139
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Graduation Range
    Availability
    Price
  • 30 ml
    5 - 30 ml
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    213-7130
    Environmentally Preferable
    Environmental certification
  • 50 ml
    10 - 50 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 67,70 €
    67,70 €
    1 * 12 ST
       67,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7131
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 100 ml
    10 - 100 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 78,10 €
    78,10 €
    1 * 12 ST
       78,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7132
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 150 ml
    20 - 150 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 94,70 €
    94,70 €
    1 * 12 ST
       94,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7133
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 250 ml
    25 - 250 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 59,30 €
    59,30 €
    1 * 6 ST
       59,30 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7134
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 400 ml
    50 - 400 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 84,90 €
    84,90 €
    1 * 6 ST
       84,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7135
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 600 ml
    100 - 600 ml
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 73,40 €
    73,40 €
    1 * 4 ST
       73,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7136
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 1000 ml
    100 - 1000 ml
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 65,70 €
    65,70 €
    1 * 3 ST
       65,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7137
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 2000 ml
    200 - 2000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,90 €
    52,90 €
    1 * 1 ST
       52,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7138
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 4000 ml
    500 - 4000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,00 €
    75,00 €
    1 * 1 ST
       75,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    213-7139
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603)
    NAR Certificate of Retirement
      Low Manufacturing Impact