Orders LinkContactUsLinkComponent
Wire baskets
Supplier:   BOCHEM
Wire baskets
Supplier:  BOCHEM
Certificates
Wire baskets
8 Options Available Below
Product Details & Documents
Stainless steel.
  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Stowable handle
These round baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
Product Family Options
Product Information
  • Ø×H
    Availability
    Price
  • 100×80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,40 €
    68,40 €
    1 * 1 ST
       68,40 €
  • 120×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,90 €
    70,90 €
    1 * 1 ST
       70,90 €
  • 150×120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,00 €
    84,00 €
    1 * 1 ST
       84,00 €
  • 180×160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • 210×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 128,00 €
    128,00 €
    1 * 1 ST
       128,00 €
  • 240×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 154,00 €
    154,00 €
    1 * 1 ST
       154,00 €
  • 270×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 172,00 €
    172,00 €
    1 * 1 ST
       172,00 €
  • 300×200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,00 €
    196,00 €
    1 * 1 ST
       196,00 €