ContactUsLinkComponent
Twist ties, SP Bel-Art
  129-1025
undefined
Twist ties, SP Bel-Art
  129-1025
  • Colour:
    Green
  • Description:
    Twist ties
  • Dimensions:
    5 mm×457,20 m
  • Pk:
    1 Roll
Strong twist tie bag closures easier to use than string or rubber bands. Made of soft, copper-tinned, annealed iron wire sealed in extruded virgin vinyl.
  • Cut the length required and twist the ends together
  • For temperature ranges from −45 to +121 °C
Ideal to bundle hard-to-handle items like rods, wires or tubes.