ContactUsLinkComponent
Label removal spray, Solvent 50
Label removal spray, Solvent 50
Catalog # 291-1204
undefined
Label removal spray, Solvent 50
Catalog # 291-1204
Specifications
  • Description:
    Label remover, spray bottle, 200 ml
  • Pk:
    200 mL
Specifications
About this item
Label remover for hassle-free removal of paper labels.
  • After an exposure time of just two to four minutes, most labels can be removed in one go
  • Removes labels from metal, glass, cardboard and many plastics (do not use on materials that are sensitive to solvents such as plexiglass or CD cases)
  • Does not contain any CFCs