Orders LinkContactUsLinkComponent
Adapters, delivery, bent
Adapters, delivery, bent
Supplier:   Lenz Laborglas
Adapters, delivery, bent
Supplier:  Lenz Laborglas
Certificates
Adapters, delivery, bent
Adapters, delivery, bent
4 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With core and jacket. Inclined at an angle of 105°. Cone with drip point.
Documents
Product Family Options
Product Information
  • Socket size
    Cone size
    Availability
    Price
  • 14/23 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,30 €
    21,30 €
    1 * 1 ST
       21,30 €
  • 19/26 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,60 €
    23,60 €
    1 * 1 ST
       23,60 €
  • 24/29 NS
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,70 €
    36,70 €
    1 * 1 ST
       36,70 €
  • 29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,90 €
    28,90 €
    1 * 1 ST
       28,90 €