Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
Supplier:   Lenz Laborglas
Multiple adapters
Supplier:  Lenz Laborglas
Certificates
Multiple adapters
Multiple adapters
4 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two standard ground 29/32 sockets and one standard ground 29/32 cone. Side neck inclined upwards at an angle of 105°.
Documents
Product Family Options
Product Information
  • Cone size
    Socket size
    Availability
    Price
  • 14/23 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,80 €
    30,80 €
    1 * 1 ST
       30,80 €
  • 19/26 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,50 €
    33,50 €
    1 * 1 ST
       33,50 €
  • 29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,40 €
    36,40 €
    1 * 1 ST
       36,40 €
  • 29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,90 €
    43,90 €
    1 * 1 ST
       43,90 €