Orders LinkContactUsLinkComponent
Adapters, cone/screw thread, Quickfit®
Adapters, cone/screw thread, Quickfit®
Supplier:   SciLabware
Adapters, cone/screw thread, Quickfit®
Supplier:  SciLabware
Certificates
Adapters, cone/screw thread, Quickfit®
Adapters, cone/screw thread, Quickfit®
11 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer. Ideal for securing thermometers, gas inlet tubes etc.
Product Family Options
Product Information
  • Cone size
    Thread
    Hole Ø
    Availability
    Price
  • 14/23 NS
    13 mm
    6,0 - 7,0 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 68,50 €
    68,50 €
    1 * 2 ST
       68,50 €
  • 14/23 NS
    18 mm
    7,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 74,30 €
    74,30 €
    1 * 2 ST
       74,30 €
  • 19/26 NS
    13 mm
    6,0 - 7,0 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 68,50 €
    68,50 €
    1 * 2 ST
       68,50 €
  • 19/26 NS
    18 mm
    7,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 74,30 €
    74,30 €
    1 * 2 ST
       74,30 €
  • 19/26 NS
    24 mm
    10,5 - 11,5 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 91,80 €
    91,80 €
    1 * 2 ST
       91,80 €
  • 24/29 NS
    13 mm
    6,0 - 7,0 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 74,30 €
    74,30 €
    1 * 2 ST
       74,30 €
  • 24/29 NS
    18 mm
    7,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 74,30 €
    74,30 €
    1 * 2 ST
       74,30 €
  • 24/29 NS
    24 mm
    10,5 - 11,5 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 80,90 €
    80,90 €
    1 * 2 ST
       80,90 €
  • 29/32 NS
    13 mm
    6,0 - 7,0 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 80,90 €
    80,90 €
    1 * 2 ST
       80,90 €
  • 29/32 NS
    18 mm
    7,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 80,90 €
    80,90 €
    1 * 2 ST
       80,90 €
  • 29/32 NS
    24 mm
    10,5 - 11,5 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 91,80 €
    91,80 €
    1 * 2 ST
       91,80 €