ContactUsLinkComponent
Optical bottom plates, 96-well, CC²™ coated, Nunc™
Optical bottom plates, 96-well, CC²™ coated, Nunc™
  734-1354
   Low Manufacturing Impact
undefined
Optical bottom plates, 96-well, CC²™ coated, Nunc™
  734-1354
   Low Manufacturing Impact
  • No. of wells:
    96
  • Recommended working volume:
    50 - 200 µl/well
  • Sterile:
    +
  • Colour:
    Black
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    30
  • Well volume:
    400 µl
PS with coverglass base, sterile, with lid. CC2™ surface treatment closely mimics a biological surface similar to Poly-D-Lysine and is a superior surface for attachment and growth of fastidious cells.
  • White or black upper structure with coverglass bottom for minimum light scatter and low auto-fluorescence, ensuring accurate results due to higher signal-to-noise ratios
  • Optimum clarity for viewing well contents