Orders LinkContactUsLinkComponent
Tanks, rectangular, with cover, Nalgene®
Tanks, rectangular, with cover, Nalgene®
   Sustainable Options Available
Tanks, rectangular, with cover, Nalgene®
   Sustainable Options Available
Certificates
Tanks, rectangular, with cover, Nalgene®
7 Options Available Below
Product Details & Documents
These heavy-duty rectangular LLDPE tanks with cover can withstand most acid, alcohols and bases. Tanks feature stepped flange at top provides reinforcement and grip for lifting.
  • Impact- and abrasion-resistant
  • Molded as one piece, no seams
  • Nominal wall thickness is 3,97 mm
  • Stepped flange at top provides reinforcement and grip for lifting
  • Not autoclavable
Specifications for Product Family
Specifications
  • Catalog No:
  • 216-1912
  • 216-1913
  • 216-1914
  • 216-1915
  • NALG14100-0040
  • NALG14100-0045
  • NALG14100-0065
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • 20 L
    360×250×250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 456,00 €
    456,00 €
    1 * 1 ST
       456,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-1912
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 30 L
    300×300×300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 277,00 €
    277,00 €
    1 * 1 ST
       277,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-1913
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 38 L
    450×300×300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 365,00 €
    365,00 €
    1 * 1 ST
       365,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-1914
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 50 L
    610×300×300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 466,00 €
    466,00 €
    1 * 1 ST
       466,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-1915
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 60 L
    450×300×450 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 453,00 €
    453,00 €
    1 * 1 ST
       453,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG14100-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 120 L
    610×450×450 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 597,00 €
    597,00 €
    1 * 1 ST
       597,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG14100-0045
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 160 L
    610×450×610 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 801,00 €
    801,00 €
    1 * 1 ST
       801,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG14100-0065
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact