Order Entry
Belgium
ContactUsLinkComponent
Sodium sulphate, tablets, KJELTABS NA mineralisation catalyst
Sodium sulphate, tablets, KJELTABS NA mineralisation catalyst
Catalog # THOMAA40
Supplier:  THOMPSON & CAPPER
CAS Number:  
undefined
Sodium sulphate, tablets, KJELTABS NA mineralisation catalyst
Catalog # THOMAA40
Supplier:  THOMPSON & CAPPER
CAS Number:  

Specifications

  • Pk:
    1.000 Tab.
  • Seq:
    0980
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt
 :
Merck Index:12,08829
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:2.7 g/cm³ (20 °C)
 :142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
CAS Number:7757-82-6
Formula:Na₂SO₄
MDL Number:MFCD00003504