ContactUsLinkComponent
pH indicator paper, phenolphthalein, Whatman™
pH indicator paper, phenolphthalein, Whatman™
Catalog # 700-9759
undefined
pH indicator paper, phenolphthalein, Whatman™
Catalog # 700-9759
Specifications
  • pH range:
    8,3 - 10
  • Description:
    Acid alkali litmus paper dispenser reel, phenolphthalein test paper, 7 mm×5 m
  • Pk:
    1 Roll
Specifications
About this item
Reel phenophthalein standard litmus paper.
  • White paper changes to pink at pH 8,3 and becomes red at pH 10
Used to determine the neutralization point in weak acid/strong alkali reactions.
Packaging: Packed as single roll.