Tritolyl phosphate (mixture of isomers), Technical Grade residue other tolyl phosphate isomers
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1330-78-5
:
Formula:C₂₁H₂₁O₄P
MDL Number:MFCD02102116
:368.37 g/mol
Merck Index:12,09892
Boiling Pt:265 °C (10 mmHg)
Melting Pt:–30 °C
Density:1.16 g/cm³
Flash Pt:234 °C
Storage Temperature:Ambient
EINECS:215-548-8
UN:2574
ADR:6.1,II
:GHS08
:GHS09
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5550 to 1.5600 (20°C, 589 nm)
- GC:63 to 85 % (sum of Tritolyl phosphate-isomers)
Specification Test Results
Frequently Bought Together