Order Entry
Belgium
ContactUsLinkComponent
Carboxymethyl cellulose sodium salt, 800 - 3100 mPas (2%, H₂O, 25°C), white solid, high viscosity, Millipore®
Carboxymethyl cellulose sodium salt, 800 - 3100 mPas (2%, H₂O, 25°C), white solid, high viscosity, Millipore®
Catalog # 217274-250
CAS Number:  
Carboxymethyl cellulose sodium salt, 800 - 3100 mPas (2%, H₂O, 25°C), white solid, high viscosity, Millipore®
Catalog # 217274-250
CAS Number:  

Specifications

  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:9004-32-4
Formula:[C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number:MFCD00081472