Order Entry
Belgium
Orders LinkContactUsLinkComponent
Anthralin

Anthralin

Glass bottle for solids, 1 g
Catalog # MOLE10673396-1G
Marketsource
Supplier:   Molekula
Anthralin
Catalog # MOLE10673396-1G
Supplier:  Molekula
Marketsource
CAS Number:  1143-38-0
Anthralin

Glass bottle for solids, 1 g

CAS Number:  1143-38-0
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  1,8,9-Trihydroxyanthracene, 1,8,9-Anthracenetriol, DIT, Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-one
  • CAS Number:1143-38-0
  • Formula:C₁₄H₁₀O₃
  • MDL Number:MFCD00053409
  • Molecular Weight:226.23 g/mol
  • Merck Index:12,00723
  • Boiling Pt:400 °C (1013 hPa)
  • Melting Pt:193.5…195 °C
  • Density:1.433 g/cm³ (20 °C)
  • EINECS:214-538-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications