Anthralin
Glass bottle for solids, 500 mg
Catalog # PROOMM0137.00
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: 1,8,9-Trihydroxyanthracene, 1,8,9-Anthracenetriol, DIT, Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-one
- CAS Number:1143-38-0
- Formula:C₁₄H₁₀O₃
- MDL Number:MFCD00053409
- Molecular Weight:226.23 g/mol
- Merck Index:12,00723
- Boiling Pt:400 °C (1013 hPa)
- Melting Pt:193.5…195 °C
- Density:1.433 g/cm³ (20 °C)
- EINECS:214-538-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



