
Alizarine
Catalog # ROTH4476.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
Specifications
About this item
:
:240.22 g/mol
Merck Index:13,00246
Boiling Pt:430 °C (1013 hPa)
Melting Pt:290 °C
Density:1.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-782-5
:GHS07
CAS Number:72-48-0
Formula:C₁₄H₈O₄
MDL Number:MFCD00001201