Trimethyl orthoacetate
MOLE26204097-100G
: Molekula
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
:
:120.15 g/mol
:109 °C (1013 hPa)
:–58 °C
:0.944 g/cm³ (20 °C)
:15 °C
:215-892-9
:3272
:3,II
:1445-45-0
:C₅H₁₂O₃
:MFCD00008477