Order Entry
Belgium
ContactUsLinkComponent
Tuning solution 2 in dil. nitric acid ICP-MS-TUNSOL2 for ICP-MS
Tuning solution 2 in dil. nitric acid ICP-MS-TUNSOL2 for ICP-MS
Catalog # ACCUICPMSTUNSOL21
Supplier:  AccuStandard
CAS Number:  
Tuning solution 2 in dil. nitric acid ICP-MS-TUNSOL2 for ICP-MS
Catalog # ACCUICPMSTUNSOL21
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Tuning solution 2 in dil. nitric acid ICP-MS-TUNSOL2 for ICP-MS

Specifications

About this item