Order Entry
Belgium
ContactUsLinkComponent
Acetic acid 0.1 mol/l (0.1 N) volumetric solution
Acetic acid 0.1 mol/l (0.1 N) volumetric solution
Catalog # REGECH20101
Supplier:  REAGECON
CAS Number:  
undefined
Acetic acid 0.1 mol/l (0.1 N) volumetric solution
Catalog # REGECH20101
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152