Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dioxane
1,4-Dioxane 0.01% in aqueous solution Reag. Ph. Eur.
1,4-Dioxane 0.01% in aqueous solution Reag. Ph. Eur.
87822.180
:
CUSTOM MADE CHEMICALS LAB
:
1,4-Dioxane 0.01% in aqueous solution Reag. Ph. Eur.
87822.180
:
CUSTOM MADE CHEMICALS LAB
:
Pack type:
Plastic bottle
Pk:
100 mL
Seq:
2440
:
:
88.11 g/mol
:
204-661-8
:
1165
:
3,II
:
123-91-1
:
C₄H₈O₂
:
MFCD00006571