Dissolved oxygen probes
Supplier: Mettler - Toledo
Jump to:
About this item
4 Options Available Below
Product Details & Documents
Documents
Mettler InLab® Sensors
Product Literature
Product Family Options
Product Information
- TypeConcentrationTemp.MaterialConnectionØ×LAvailabilityPrice
- 664-1004Dissolved oxygen probe InLab® 605, IP 67Dissolved oxygen probe InLab® 605, IP 67Dissolved oxygen probe InLab® 605, IP 670 - 200%, 0 - 20 mg/l0...+60 °CPPSBNC/RCA (Cinch), 1,8 m cable12×120 mmLog in for availability1 * 1 ST1 060,00 €
- 664-1005Dissolved oxygen probe InLab® 605, IP 67Dissolved oxygen probe InLab® 605, IP 67Dissolved oxygen probe InLab® 605, IP 670 - 200%, 0 - 20 mg/l0...+60 °CPPSBNC/RCA (Cinch), 10 m cable12×120 mmLog in for availability1 * 1 ST1 220,00 €
- 664-0204Dissolved oxygen probe LE621, galvanic, with NTC30 KΩ temperature probeDissolved oxygen probe LE621, galvanic, with NTC30 KΩ temperature probeLE621, galvanic, with NTC30 KΩ0 - 200%, 0 - 20 mg/l0...+50 °CPVCBNC, 1 m cable12×120 mmLog in for availability1 * 1 ST467,00 €
- 664-0217Dissolved oxygen probe LE621 (IP 67), galvanic, with NTC30 KΩ temperature probeDissolved oxygen probe LE621 (IP 67), galvanic, with NTC30 KΩ temperature probeLE621 (IP 67), galvanic, with NTC30 KΩ0 - 200%, 0 - 20 mg/l0...+50 °CPVCBNC/Cinch, 1 m cable12×120 mmLog in for availability1 * 1 ST499,00 €
Accessories
Product Information
- Catalog #AvailabilityPrice
- Electrolyte for InLab® 605 DO probe, one 25 ml bottleElectrolyte for InLab® 605 DO probe, one 25 ml bottleMETT51340294Log in for availability1 * 25 mL - 30,20 €
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for InLab® 605 DO probe, incl. three membrane bodies and 25 ml of electrolyteMembrane kit for InLab® 605 DO probe, incl. three membrane bodies and 25 ml of electrolyte664-1006Log in for availability1 * 1 ST - 304,00 €
Product Information
- Catalog #AvailabilityPrice
- Calibration bottle for InLab® 605 DO probe
Product Information
- Catalog #AvailabilityPrice
- Sealing kit for InLab® 605 DO probe
Product Information
- Catalog #AvailabilityPrice
- Zero oxygen tablets, used for calibration