Orders LinkContactUsLinkComponent
Frocks, PP
Frocks, PP
Supplier:   Medicom
Frocks, PP
Supplier:  Medicom
Certificates
Frocks, PP
4 Options Available Below
Product Details & Documents
These single-use hygiene frocks made from PP protect the working environment from particles coming from the operator's body (dander, hair, perspiration, etc.).
  • With collar and long sleeves
  • Elasticated cuffs
  • Snap buttons
  • Without pockets
Weight: 40 g/m²
Specifications for Product Family
Specifications
  • Catalog No:
  • HOPE40.301/04
  • HOPE40.301/05
  • HOPE40.301V/04
  • HOPE40.301V/05
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Garment application:
  • General Purpose|Industrial
  • Industrial|General Purpose
  • Industrial|General Purpose
  • General Purpose|Industrial
  • Weight:
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • White
    XL
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 74,50 €
    74,50 €
    1 * 50 ST
       74,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    HOPE40.301/04
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Garment application
    General Purpose|Industrial
    Weight
    40 g/m²
  • White
    XXL
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 74,50 €
    74,50 €
    1 * 50 ST
       74,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    HOPE40.301/05
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Garment application
    Industrial|General Purpose
    Weight
    40 g/m²
  • Green
    XL
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 74,50 €
    74,50 €
    1 * 50 ST
       74,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    HOPE40.301V/04
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Garment application
    Industrial|General Purpose
    Weight
    40 g/m²
  • Green
    XXL
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 74,50 €
    74,50 €
    1 * 50 ST
       74,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    HOPE40.301V/05
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Garment application
    General Purpose|Industrial
    Weight
    40 g/m²