Order Entry
Belgium
ContactUsLinkComponent
Mouse Recombinant HSC Expansion PB (from E. coli)
Mouse Recombinant HSC Expansion PB (from E. coli)
Catalog # SHBTPB-500-04
New Product
CAS Number:  
Mouse Recombinant HSC Expansion PB (from E. coli)
Catalog # SHBTPB-500-04
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 KIT
  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Seq:
    0075
  • Storage conditions:
    At or below −20 °C
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Endotoxin level:
    Low
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

About this item

Cytokines for polarizing T cells into T regulatory cells. T regulatory cells (Tregs) are a subset of T cells that suppress immune function. They are characterized by the expression of CD4, CD25 and Foxp3 and are known to produce immunosuppressive cytokines TGF-β 1 and IL-10. Tregs can be cultured from naive CD4+ T cells by culturing them in the presence of IL-2 and TGF-β 1 with CD3 and CD28 stimulation. In some cases cells are also cultured with neutralizing antibodies to IL-4, IFNγ and IL-12 and/or retinoic acid.

  • High quality
  • Low endotoxin
  • Affordable

Studies have shown that decreased expression of CD34 tends to correlate with decreased self renewal properties and pluripotent potential. Culturing CD34+ cells in a cytokine mixture of TPO, SCF, and FLT-3 Ligand for short time periods allows CD34+ cells to proliferate without decreasing CD34 expression, thus retaining their stem cell properties.

Caution: For research use only.