Order Entry
Belgium
ContactUsLinkComponent
 
Human Recombinant IL-21 (CTGrade GMP) (from E. coli)
New Product
 
Human Recombinant IL-21 (CTGrade GMP) (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    At or below −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.05
  • Biological activity:
    Bioactivity assay: Proliferation of B9 cell, ED50 ≤5 ng/ml, bioactivity ≥WHO standard not available
  • Gene ID:
    Q9HBE4
  • Reconstitution instructions:
    Sterile water at 0,1 - 0,5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Purity:
    ≥97%
  • Molecular weight:
    15,4 kDa
  • Sequence:
    MQDRHMIRMR QLIDIVDQLK NYVNDLVPEF LPAPEDVETN CEWSAFSCFQ KAQLKSANTG NNERIINVSI KKLKRKPPST NAGRRQKHRL TCPSCDSYEK KPPKEFLERF KSLLQKMIHQ HLSSRTHGSE DS
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    GMP
  • Shipping temperature:
    Ambient
  • Tested applications:
    Bioassay

 

Frequently Bought Together