Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant TH17 Cell Polarizing Plus PB (from E. coli)
Human Recombinant TH17 Cell Polarizing Plus PB (from E. coli)
Catalog # SHBTPB-500-12
New Product
CAS Number:  
Human Recombinant TH17 Cell Polarizing Plus PB (from E. coli)
Catalog # SHBTPB-500-12
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 KIT
  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    0075
  • Storage conditions:
    At or below −20 °C
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Endotoxin level:
    Low
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

About this item

Cytokines for polarizing human T cells into Th9 cells. Th9 cells are a unique subset of T helper cells that produce IL-9. An important function of IL-9 producing T cells is fighting inflammation and helminth infections. Th9 cells can be generated from naive CD4+ T cells in culture by culturing them in the presence of IL-4 and TGF-β 1. It also appears that IFNγ and IL-27 can inhibit the generation of Th9 cells.

  • High quality
  • Low endotoxin
  • Affordable

Th17 cells are characterized by their ability to produce IL-17, IL-22 and IL-21. Th17 cells can be cultured from naive CD4+ T cells by culturing them in the presence of anti-CD3 and anti-CD28 stimulation with recombinant IL-6 and recombinant TGF-β 1. In some cases neutralizing antibodies against IL-12, IL-4 and IFNγ are used along with recombinat cytokines IL-1 β, IL-21 and/or IL-23.

Caution: For research use only.