Order Entry
Belgium
ContactUsLinkComponent
Mouse Recombinant Mouse Glioma CultuPB (from E. coli)
Mouse Recombinant Mouse Glioma CultuPB (from E. coli)
Catalog # SHBTPB-500-20
New Product
CAS Number:  
Mouse Recombinant Mouse Glioma CultuPB (from E. coli)
Catalog # SHBTPB-500-20
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 KIT
  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Seq:
    0075
  • Storage conditions:
    At or below –20 °C
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein/peptide name:
    Mouse Glioma CultuPB
  • Sequence:
    NA
  • Endotoxin level:
    Low
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

About this item

Cytokines for cell culture of mouse glioma cells. A glioma is a generic term used to describe tumors that arise from the glia tissue of the brain that supports and surrounds neurons. Glioma tumor cells can be successfully cultured ex vivo with EGF, FGF-basic (or FGF-2), and PDGFs (AA and BB).

  • High quality
  • Low endotoxin
  • Affordable
Delivery: 2×100 μg of Recombinant mouse EGF, 200 μg of Recombinant mouse FGF-basic/FGF-2, 100 μg of Recombinant mouse PDGF-AA and 100 μg of Recombinant mouse PDGF-BB.
Caution: For research use only.