Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant SCF (CTGrade GMP) (from E. coli)
Human Recombinant SCF (CTGrade GMP) (from E. coli)
New Product
Certificates
Human Recombinant SCF (CTGrade GMP) (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,05
  • Biological activity:
    Bioactivity Assay: Proliferation of TF-1, ED₅₀ ≤15 ng/ml, Bioactivity ≥1,0x10³ IU/µg
  • Gene ID:
    P21583
  • Reconstitution instructions:
    Sterile water at 0,1 - 0,5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    c-Kit Ligand|KL|mgF|Steel Factor
  • Protein/peptide name:
    CTGrade GMP SCF
  • Purity:
    ≥97%
  • Molecular weight:
    18,6 kDa
  • Sequence:
    MEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVA
  • Endotoxin level:
    Low
  • Formulation:
    10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    GMP
  • Shipping temperature:
    Ambient
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together