Order Entry
Belgium
ContactUsLinkComponent
 
Horse Recombinant PDGF-BB (from E. coli)
New Product
 
Horse Recombinant PDGF-BB (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Horse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity Assay: NR6R 3T3 proliferation, ED₅₀ ≤30 ng/ml, Bioactivity ≥3,3×10⁴ units/mg
  • Gene ID:
    XP_005606703
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    ODGF|GDGF
  • Protein/peptide name:
    PDGF-BB
  • Purity:
    ≥95%
  • Molecular weight:
    24,8 kDa
  • Sequence:
    MSLGSLAVAE PAMIAECKTR TEVFEISRRL IDRTNANFLV WPPCVEVQRC SGCCNNRHVQ CRPTQVQLRP VQVRKIEIVR KKPTFKKATV TLEDHLACKC ETVGAARPVT
  • Endotoxin level:
    Low
  • Formulation:
    10 mM Sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together