Order Entry
Belgium
ContactUsLinkComponent
Sterivex™ Membrane Filter Units, with Durapore® (PVDF) Membrane
Sterivex™ Membrane Filter Units, with Durapore® (PVDF) Membrane
Supplier:  Merck
Certificates
undefined
Sterivex™ Membrane Filter Units, with Durapore® (PVDF) Membrane
Supplier:  Merck

Specifications

  • Catalog No:
  • MILFSVGVL10RC
  • MILFSVGV010RS
  • MILFSVHV010RS
  • MILFSVGVB1010
  • MILFSVHVL10RC
  • MILFSVHVB1010
  • MILFSVGV01015
  • MILFSVHV01015
  • Filters:
  • Durapore® (PVDF), 0,22 µm, hydrophilic
  • Durapore® (PVDF), 0,22 µm, hydrophilic
  • Durapore® (PVDF), 0,45 µm, hydrophilic
  • Durapore® (PVDF), 0,22 µm, hydrophilic
  • Durapore® (PVDF), 0,45 µm, hydrophilic
  • Durapore® (PVDF), 0,45 µm, hydrophilic
  • Durapore® (PVDF), 0,22 µm, hydrophilic
  • Durapore® (PVDF), 0,45 µm, hydrophilic
  • Capacity:
  • 1000 ml
  • 1000 ml
  • 1000 ml
  • 1000 ml
  • 1000 ml
  • 1000 ml
  • 1000 ml
  • 1000 ml
  • Filtration area:
  • 10 cm²
  • 10 cm²
  • 10 cm²
  • 10 cm²
  • 10 cm²
  • 10 cm²
  • 10 cm²
  • 10 cm²
  • Max. temperature:
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C

Specifications

Related Information

Frequently Bought Together