ContactUsLinkComponent
Millicell® Cell Culture Inserts, Hydrophilic PTFE (Biopore™) Membrane
Millicell® Cell Culture Inserts, Hydrophilic PTFE (Biopore™) Membrane
Supplier:  Merck
Certificates
undefined
Millicell® Cell Culture Inserts, Hydrophilic PTFE (Biopore™) Membrane
Supplier:  Merck
Specifications
  • Catalog No:
  • MILFPICM01250
  • MILFPICM0RG50
  • MILFPICM03050
  • Effective diameter of membrane:
  • 12 mm
  • 30 mm
  • 30 mm
  • Overall height:
  • 10,5 mm
  • 5 mm
  • 13 mm