Millex® Syringe Filters, HPF, Durapore (PVDF) Membrane with Glass Fibre Pre-filter, Merck
Fournisseur: Merck
Certificats
À propos de cet article
These filters include multiple media: a graduated prefilter and a membrane filter for high throughput filtration of viscous or particle-laden solutions.
- High-density polyethylene housing compatible with the most commonly used solvents
- Multi-layer filter can improve throughput of high particulate/viscous samples
Applications include analytical sample preparation, wine analysis and general particulate removal.
Frequently Bought Together