Order Entry
Belgium
Orders LinkContactUsLinkComponent
Naval Patch Test Kit

Naval Patch Test Kit

Naval patch test kit, 1
Catalog # MILFXX6504700
Supplier:   Merck
Naval Patch Test Kit
Catalog # MILFXX6504700
Supplier:  Merck
CAS Number:  
Naval Patch Test Kit
Naval Patch Test Kit

Naval patch test kit, 1

CAS Number:  
Price
5 480,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Patch Test Kit is a complete solution for field-based collection and analysis of hydrocarbon-based hydraulic fluids, bulk chemicals, boiler water, and lubricating oils.

  • Monitors particulate contamination rapidly in hydrocarbon-based hydraulic fluids, bulk chemicals, boiler water and lubricating oils
  • Filter color rating and particle assessment scales correspond to recognised standard contamination levels
  • Detects significant changes in cleanliness through dependable and sensitive test results
  • Comes with filters for 100 tests

It allows the detection of significant changes in cleanliness through dependable, sensitive, colorimetric-based analysis.

Features:
- Contains enough expendable materials for 100 tests
- Contained within a lightweight, easy-to-carry case
- Filter color rating and particle assessment scales correspond to recognized standard contamination levels

Delivery information: Kit contains: MILFPD1504700 PetriSlides for contamination analysis, MILFSMWP02500 MF-Millipore Membrane Filter, 5,0 µm, 25 mm, white, plain, 100, MILFSMWP04700 MF-Millipore Membrane Filter, 5,0 µm, 47 mm, white, plain, 100, MILFXX6200006P Filter forceps, blunt end, stainless steel, MILFXX6200035 Hand Vacuum Pump, stainless steel, MILFXX6300120 Patch Test Filter Holder Assembly, MILFXX6504704 Solvent Dispensing Bottle, MILFXX6504707 Swinnex 25 mm with back-pressure screen, MILFXX6504709 PVC Bottle, 120 ml, MILFXX6504710 Tubing for vacuum, MILFXX6504713 Naval Fluid Color Guide 1.

Specifications

Specifications

  • Pk:
    1
  • Description:
    Naval patch test kit

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.