Microfil® Funnel and Filters
Leverancier: Merck
Certificaten
Over dit artikel
Microfil® funnels and filters assuring leak-free operation and uniform microorganism recoveries as well as an easy transfer of the membrane with forceps and prevents accidentally touching the filtration area.
- Filter color: White
- Filter surface: Gridded
- Chemistry: Mixed Cellulose Esters (MCE)
- Sterilised
Application: Specific test regulations (clinical, cosmetics, environmental, food and beverage, industrial QC).
Frequently Bought Together