Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multiscreen® Filter Plates, Opaque

Multiscreen® Filter Plates, Opaque

Fournisseur:   Merck
Multiscreen® Filter Plates, Opaque
Fournisseur:  Merck
Certificates
Multiscreen® Filter Plates, Opaque
Multiscreen® Filter Plates, Opaque

2 Options Available Below

Product Details & Documents

Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated or particular laden samples. They are also widely used for clarification of aqueous solutions.

  • Multiple configurations and applications
  • Chemical compatibility
  • Automation-compatible
  • Superior vacuum filtration and filtrate collection
  • Hydrophilic lid
  • Colour code: Opaque

Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated or particulate-laden samples. They are also widely used for clarification of aqueous solutions.

Application:
Receptor/ligand binding assays, sample preparation, TCA precipitation of protein, DNA purification.
Research category and general filtration.
Nucleic acid and assays.

Binding type: High binding surface
Height: 14,4 mm
Length: 128 mm
Width: 85,5 mm

Product Family Options

Product Information

  • Material
    Colour
    Well volume
    Membrane
    Pore size
    No. of wells
    Sterile
    Application
    Disponibilité
    Prix
  • 736-0179
    GFB plate, filtration area 0,26 cm²
    GFB plate, filtration area 0,26 cm²
    Titanium
    Opaque
    50 - 250 μl
    PVDF
    1,0 µm
    96
    -
    Receptor/ligand binding assays, sample preparation, enzyme assays with precipitation, DNA purification
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 1 850,00 €
    1 850,00 €
    1 * 50 ST
       1 850,00 €

  • MILFMAFCN0B50
    GFC plate, filtration area 0,26 cm²
    GFC plate, filtration area 0,26 cm²
    Titanium
    Opaque
    50 - 250 μl
    PVDF
    96
    -
    Receptor/Ligand Binding Assays, sample Preparation, TCA precipitation of protein
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 1 850,00 €
    1 850,00 €
    1 * 50 ST
       1 850,00 €