Orders LinkContactUsLinkComponent
Fluid Sampling Kit
Fluid sampling kit, non sterile, contains PTFE hose(s), polystyrene monitor(s), L×W×H: 368×235×125 mm, Stainless steel
Catalog # MILFXX6403730
Supplier:   Merck
Fluid Sampling Kit
Catalog # MILFXX6403730
Supplier:  Merck
Fluid Sampling Kit
Fluid Sampling Kit
Fluid sampling kit, non sterile, contains PTFE hose(s), polystyrene monitor(s), L×W×H: 368×235×125 mm, Stainless steel
Price
4 690,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Fluid sampling kit for sampling turbine fuels, water or other liquids through Millipore contamination analysis monitors.
  • Sturdy carrying case and stainless steel sampling assembly provide years of in-field service
  • Fluid Sampling Kit includes 12 Tenite contamination monitors
The Fluid Sampling Kit is a complete solution for field-based fluid collection and contamination analysis. The stainless steel sampler assembly plugs into a quick-release valve installed in the system line, which allows a measured quantity of liquid to pass through the monitor. A valved syringe serves as a pump to remove residual liquid from the monitor. Kit is compatible with our 37 mm contamination monitors and matched-weight monitors.
Specifications
  • Description:
    Fluid sampling kit, non sterile, contains PTFE hose(s), polystyrene monitor(s), L×W×H: 368×235×125 mm
  • Material:
    Stainless steel
Accessories
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Colour standards
    Colour standards
    MILFASTM03701
    1
    Log in for availability
    1 * 1 ST - 249,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Filter holder gasket, PTFE
    Filter holder gasket, PTFE
    MILFXX3002502
    Log in for availability
    1 * 10 ST - 105,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Filter holder support screen, stainless steel
    Filter holder support screen, stainless steel
    MILFXX3002510
    Log in for availability
    1 * 1 ST - 56,50 €