Order Entry
Belgium
Orders LinkContactUsLinkComponent
Patch Test Filter Holder Assembly

Patch Test Filter Holder Assembly

Patch test filter holder assembly
Catalog # MILFXX6300120
Supplier:   Merck
Patch Test Filter Holder Assembly
Catalog # MILFXX6300120
Supplier:  Merck
Patch Test Filter Holder Assembly
Patch Test Filter Holder Assembly

Patch test filter holder assembly

Price
2 300,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Patch Test Kit is a complete solution for field-based collection and analysis of hydrocarbon-based hydraulic fluids, bulk chemicals, boiler water, and lubricating oils.

  • Monitors particulate contamination rapidly in hydrocarbon-based hydraulic fluids, bulk chemicals, boiler water and lubricating oils
  • Filter colour rating and particle assessment scales correspond to recognized standard contamination levels
  • Detects significant changes in cleanliness through dependable and sensitive test results
  • Comes with filters for 100 tests

It allows the detection of significant changes in cleanliness through dependable, sensitive, colorimetric-based analysis.

Applications: In-field contamination analysis of hydrocarbon-based hydraulic fluids, water, lubricating oils and related products.

Features:
Contains enough expendable materials for 100 tests
Contained within a lightweight, easy-to-carry case
Filter color rating and particle assessment scales correspond to recognized standard contamination levels

Delivery information: Kit contains: MILFPD1504700 PetriSlides for contamination analysis, MILFSMWP02500 MF-Millipore Membrane Filter, 5,0 µm, 25 mm, white, plain, 100, MILFSMWP04700 MF-Millipore Membrane Filter, 5,0 µm, 47 mm, white, plain, 100, MILFXX6200006P Filter forceps, blunt end, stainless steel, MILFXX6200035 Hand Vacuum Pump, stainless steel, MILFXX6300120 Patch Test Filter Holder Assembly, MILFXX6504704 Solvent Dispensing Bottle, MILFXX6504707 Swinnex 25 mm with back-pressure screen, MILFXX6504709 PVC Bottle, 120 ml, MILFXX6504710 Tubing for vacuum, MILFXX6504713 Naval Fluid Color Guide 1.

Specifications
  • Description:
    Patch test filter holder assembly