Order Entry
Belgium
ContactUsLinkComponent
Upstate® Autorad Orientation Markers for Western Blotting
Upstate® Autorad Orientation Markers for Western Blotting
Catalog # MILF20-135
Supplier:  Merck
CAS Number:  
undefined
Upstate® Autorad Orientation Markers for Western Blotting
Catalog # MILF20-135
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5
  • Description:
    Autorad orientation markers
  • Shelf life:
    1 year
  • Storage temperature:
    RT

Specifications

About this item

Autorad orientation markers for use on immunoblots in detection of Chemiluminescence in Western blotting, dot/slot blotting.