Order Entry
Belgium
ContactUsLinkComponent
Solvent-Resistant Stirred Cell for 76 mm Membranes
Solvent-Resistant Stirred Cell for 76 mm Membranes
Catalog # MILFXFUF07601
Supplier:  Merck
undefined
Solvent-Resistant Stirred Cell for 76 mm Membranes
Catalog # MILFXFUF07601
Supplier:  Merck

Specifications

  • Description:
    Solvent-resistant stirred cells, for 76 mm membranes
  • Pk:
    1

Specifications

About this item

For applications requiring compatibility with solvents, this Millipore stirred cell is available in two sizes. Certain aldehydes, ketones (e.g., acetone) and aliphatic ethers/esters may reduce the life of the fluorocarbon O-rings.

  • Borosilicate glass cylinder and PTFE components for broad compatibility
  • Autoclavable with membrane in-place
  • Top plate opening provides access to contents without dismantling
  • Few components, so its easy to clean and assemble
  • For 47 mm disc filters

Applications are concentration and buffer exchange involving organic solvents.

Membrane type: Solvent resistant
Working volume: 300 ml
Ø×H: 76×165 mm
Filtration area: 40 cm²