Orders LinkContactUsLinkComponent
MultiScreen® HTS Filter Plates, Styrene
MultiScreen® HTS Filter Plates, Styrene
Supplier:   Merck
MultiScreen® HTS Filter Plates, Styrene
Supplier:  Merck
Certificates
MultiScreen® HTS Filter Plates, Styrene
2 Options Available Below
Product Details & Documents
MultiScreen plates have a long history as a reliable tool for the advancement of drug discovery and life science research.
  • Enhanced handling is achieved with rigid sidewalls
  • Abundant surface area for barcode labels
  • Prevents any cross-contamination during incubation
  • High throughput screening
  • Color code: Clear
MultiScreen HTS plates are specifically developed for high throughput use with automated workstations. Plasmid purification is a widely employed method in molecular cloning, transformation, and recombinant protein expression. Plasmid purification kits are designed to accommodate different bacterial culture sizes and the corresponding plasmid yield.
Multiscreen HTS filter plate, styrene is suitable for use in the clearing of bacterial lysates with the MultiScreen HTS vacuum manifold prior to plasmid purification using MILFMSNUPSD50.
Product Family Options
Product Information
  • Material
    Well volume
    No. of wells
    Sterile
    Availability
    Price
  • MILFMSNANLY10
    Multiscreen HTS filter plate
    Multiscreen HTS filter plate
    Polystyrene
    50 - 250 μl
    96
    -
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST
  • MILFMSNANLY50
    Multiscreen HTS filter plate
    Multiscreen HTS filter plate
    Polystyrene
    50 - 250 μl
    96
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST