Orders LinkContactUsLinkComponent
MultiScreen® PCR 96 Filter Plates
MultiScreen® PCR 96 Filter Plates
Supplier:   Merck
MultiScreen® PCR 96 Filter Plates
Supplier:  Merck
Certificates
MultiScreen® PCR 96 Filter Plates
MultiScreen® PCR 96 Filter Plates
2 Options Available Below
Product Details & Documents
The polymerase chain reaction (PCR) is a highly effective fundamental technique in molecular biology. It offers a fast and efficient way to amplify specific DNA or RNA sequences from nucleic acids derived from different sources using enzymatic processes in a laboratory setting.
  • Application: Sample preparation
  • Binding type: Low binding surface
  • Ambient
We have an extensive range of PCR consumables and equipment designed to facilitate every stage of your PCR workflow MultiScreen-PCR 96 Filter plates can be used to purify PCR products to be used in further downstream applications.
MultiScreen-PCR 96 filter plate is suitable for use:
-In receptor/ligand binding assays
-In sample preparation
-In enzyme assays with precipitation
-In DNA purification
-In polymerase chain reaction (PCR)
-In purifying polymerase chain reaction (PCR) products for mutation analysis
-In purifying PCR products for measuring CAG repeats
-In purifying PCR products for sequencing
Product Family Options
Product Information
  • Material
    Well volume
    Recommended working volume
    Membrane
    No. of wells
    Sterile
    Availability
    Price
  • MILFMSNU03010
    MultiScreen PCR filter plate, with lid
    MultiScreen PCR filter plate, with lid
    Polystyrene
    300 μl
    150 - 300 μl
    Proprietary
    96
    -
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST
  • MILFMSNU03050
    MultiScreen PCR filter plate, with lid
    MultiScreen PCR filter plate, with lid
    Polystyrene
    300 μl
    150 - 300 μl
    Proprietary
    96
    -
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST