Order Entry
Belgium
ContactUsLinkComponent
Multiscreen® Filter Plates with Immobilon®-P-FL membrane
Multiscreen® Filter Plates with Immobilon®-P-FL membrane
Catalog # MILFS5EJ104I07
Supplier:  Merck
Multiscreen® Filter Plates with Immobilon®-P-FL membrane
Catalog # MILFS5EJ104I07
Supplier:  Merck

Specifications

  • Application:
    Fluorescent elispot
  • Membrane:
    Immobilon®-P-FL membrane
  • No. of wells:
    96
  • Pore size:
    0,45 µm
  • Colour:
    Clear
  • Description:
    MultiScreen®HTS plates with underdrain
  • Material:
    Acrylic
  • Pk:
    10

Specifications

About this item