Orders LinkContactUsLinkComponent
Multiscreen® High Volume Filter Plates
Multiscreen® High Volume Filter Plates
Supplier:   Merck
Multiscreen® High Volume Filter Plates
Supplier:  Merck
Certificates
Multiscreen® High Volume Filter Plates
Multiscreen® High Volume Filter Plates
2 Options Available Below
Product Details & Documents
The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.
  • Sample prep
  • Protein kinase/phosphatase assays
  • Receptor/ligand binding assays
Product Family Options
Product Information
  • Material
    Colour
    Membrane
    Pore size
    No. of wells
    Sterile
    Availability
    Price
  • MILFMVHVN4525
    MultiScreen® High volume filter plates
    MultiScreen® High volume filter plates
    PP
    Opaque
    Hydrophilic Durapore®
    membrane
    0,45 µm
    96
    +
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 1 200,00 €
    1 200,00 €
    1 * 25 ST
       1 200,00 €
  • MILFMVFCN1225
    MultiScreen® High volume filter plates
    MultiScreen® High volume filter plates
    PP
    Opaque
    Glass fiber membrane
    1,2 µm
    96
    -
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 1 200,00 €
    1 200,00 €
    1 * 25 ST
       1 200,00 €